siRNA Details
Gene Symbol: Rax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053678.1 | NP_446130.1 | 2 | 436 | ||
GenBank mRNA | AF135839.1 | - | 2 | 436 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053678.1 | NP_446130.1 | 2 | 436 | ||
GenBank mRNA | AF135839.1 | - | 2 | 436 |