siRNA Details
| Gene Symbol: Slfn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053687.1 | NP_446139.1 | 6 | 2961 | ||
| XM_008767933.1 | XP_008766155.1 | 8 | 3423 | |||
| GenBank mRNA | AF168795.1 | - | 6 | 2961 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slfn3| Gene Symbol: Slfn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053687.1 | NP_446139.1 | 6 | 2961 | ||
| XM_008767933.1 | XP_008766155.1 | 8 | 3423 | |||
| GenBank mRNA | AF168795.1 | - | 6 | 2961 | ||