siRNA Details
| Gene Symbol: Cited2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053698.2 | NP_446150.1 | 2 | 596 | ||
| GenBank mRNA | AF361476.1 | - | 2 | 601 | ||
| BC087005.1 | - | 2 | 596 | 7191504 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010828, AK133791, BC057126, AK131664, Y15163, and others.

Cited2
Genomic Map