siRNA Details
Gene Symbol: Pi4k2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053735.1 | NP_446187.1 | 8, 9 | 1335 | ||
GenBank mRNA | AY029199.1 | - | 8, 9 | 1335 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pi4k2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053735.1 | NP_446187.1 | 8, 9 | 1335 | ||
GenBank mRNA | AY029199.1 | - | 8, 9 | 1335 |