siRNA Details
| Gene Symbol: TREH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301065.1 | NP_001287994.1 | 6, 7 | 695 | ||
| NM_007180.2 | NP_009111.2 | 7, 8 | 788 | |||
| GenBank mRNA | AB000824.1 | - | 7, 8 | 798 | ||
| AK091981.1 | - | 5, 6 | 657 | |||
| AK223140.1 | - | 7, 8 | 762 | |||
| AK223143.1 | - | 7, 8 | 766 | |||
| AK309565.1 | - | 7, 8 | 762 | |||
| BC109206.2 | - | 6, 7 | 689 | 40006501 | ||

TREH
Genomic Map