siRNA Details
| Gene Symbol: Kidins220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053795.1 | NP_446247.1 | 2 | 186 | ||
| XM_006239925.2 | XP_006239987.1 | 2 | 272 | |||
| XM_006239927.2 | XP_006239989.1 | 2 | 272 | |||
| XM_006239928.2 | XP_006239990.1 | 2 | 272 | |||
| XM_006239937.2 | XP_006239999.1 | 2 | 272 | |||
| GenBank mRNA | AF239045.2 | - | 2 | 186 | ||
| AF313464.1 | - | 1 | 75 | |||

Kidins220
Genomic Map