siRNA Details
| Gene Symbol: LOC102548514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235165.2 | XP_006235227.1 | 2 | 237 | ||
| Gene Symbol: Crnkl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053797.2 | NP_446249.1 | 1 | 190 | ||
| GenBank mRNA | AF245018.1 | - | 1 | 180 | ||
| BC085718.1 | - | 1 | 190 | 7190212 | ||

LOC102548514
Genomic Map