siRNA Details
| Gene Symbol: Txn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC058454.1 | - | 444 | 6886988 | ||
| FQ209943.1 | - | 441 | ||||
| FQ210839.1 | - | 444 | ||||
| FQ216849.1 | - | 470 | ||||
| FQ217822.1 | - | 439 | ||||
| FQ221646.1 | - | 412 | ||||
| X14878.1 | - | 419 | ||||

Txn1
Genomic Map