siRNA Details
| Gene Symbol: Kcnk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053806.2 | NP_446258.2 | 3 | 1283 | ||
| GenBank mRNA | AF281304.1 | - | 1283 | |||
| Gene Symbol: LOC100909725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748812.3 | XP_003748860.1 | 1810 | |||

Kcnk6
Genomic Map