siRNA Details
| Gene Symbol: Nup107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053830.2 | NP_446282.2 | 2 | 146 | ||
| GenBank mRNA | CB733157.1 | - | 2 | 157 | ||
| CB775175.1 | - | 1 | 82 | |||
| L31840.1 | - | 2 | 163 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nup107| Gene Symbol: Nup107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053830.2 | NP_446282.2 | 2 | 146 | ||
| GenBank mRNA | CB733157.1 | - | 2 | 157 | ||
| CB775175.1 | - | 1 | 82 | |||
| L31840.1 | - | 2 | 163 | |||