siRNA Details
| Gene Symbol: Nup107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053830.2 | NP_446282.2 | 20, 21 | 1800 | ||
| GenBank mRNA | CB792533.1 | - | 1, 2 | 69 | ||
| EV764995.1 | - | 747 | ||||
| L31840.1 | - | 20, 21 | 1820 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nup107| Gene Symbol: Nup107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053830.2 | NP_446282.2 | 20, 21 | 1800 | ||
| GenBank mRNA | CB792533.1 | - | 1, 2 | 69 | ||
| EV764995.1 | - | 747 | ||||
| L31840.1 | - | 20, 21 | 1820 | |||