siRNA Details
| Gene Symbol: Gucy2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053831.1 | NP_446283.1 | 2 | 516 | ||
| XM_008773386.1 | XP_008771608.1 | 1 | 259 | |||
| GenBank mRNA | L36030.1 | - | 2 | 516 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gucy2f| Gene Symbol: Gucy2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053831.1 | NP_446283.1 | 2 | 516 | ||
| XM_008773386.1 | XP_008771608.1 | 1 | 259 | |||
| GenBank mRNA | L36030.1 | - | 2 | 516 | ||