siRNA Details
| Gene Symbol: Opcml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053848.1 | NP_446300.1 | 7 | 2046 | ||
| XM_008766041.1 | XP_008764263.1 | 5 | 1204 | |||
| GenBank mRNA | M88709.1 | - | 2196 | |||
| M88710.1 | - | 7 | 1877 | |||
| M88711.1 | - | 7 | 2046 | |||

Opcml
Genomic Map