siRNA Details
| Gene Symbol: Atp6v1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053884.1 | NP_446336.1 | 2 | 473 | ||
| GenBank mRNA | U43175.1 | - | 2 | 473 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v1f| Gene Symbol: Atp6v1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053884.1 | NP_446336.1 | 2 | 473 | ||
| GenBank mRNA | U43175.1 | - | 2 | 473 | ||