siRNA Details
| Gene Symbol: Rere | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053885.2 | NP_446337.2 | 23 | 5817 | ||
| XM_006239438.2 | XP_006239500.1 | 23 | 5911 | |||
| XM_006239441.2 | XP_006239503.1 | 17 | 7821 | |||
| XM_008764198.1 | XP_008762420.1 | 23 | 5914 | |||
| XM_008764199.1 | XP_008762421.1 | 23 | 6331 | |||
| XM_008764200.1 | XP_008762422.1 | 23 | 5887 | |||
| XM_008764201.1 | XP_008762423.1 | 22 | 5789 | |||
| XM_008764202.1 | XP_008762424.1 | 22 | 5355 | |||
| XM_008764203.1 | XP_008762425.1 | 21 | 7482 | |||
| GenBank mRNA | U44091.1 | - | 22 | 6626 | ||

Rere
Genomic Map