siRNA Details
| Gene Symbol: Ppp1r12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053890.1 | NP_446342.1 | 2 | 387 | ||
| XM_006241308.1 | XP_006241370.1 | 2 | 548 | |||
| XM_006241309.1 | XP_006241371.1 | 2 | 548 | |||
| XM_006241311.1 | XP_006241373.1 | 2 | 548 | |||
| XM_006241312.1 | XP_006241374.1 | 2 | 548 | |||
| XM_006241313.1 | XP_006241375.1 | 2 | 548 | |||
| XM_006241314.1 | XP_006241376.1 | 2 | 548 | |||
| XM_006241315.1 | XP_006241377.1 | 2 | 548 | |||
| GenBank mRNA | FQ233443.1 | - | 2 | 553 | ||
| S74907.1 | - | 2 | 387 | |||
| U50185.1 | - | 2 | 267 | |||

Ppp1r12a
Genomic Map