siRNA Details
| Gene Symbol: Jdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053894.1 | NP_446346.1 | 3 | 859 | ||
| GenBank mRNA | U53449.1 | - | 3 | 859 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jdp2| Gene Symbol: Jdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053894.1 | NP_446346.1 | 3 | 859 | ||
| GenBank mRNA | U53449.1 | - | 3 | 859 | ||