siRNA Details
| Gene Symbol: Pgap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229825.2 | XP_006229887.1 | 9 | 2098 | ||
| XM_006229830.2 | XP_006229892.1 | 9 | 2087 | |||
| XM_006229831.2 | XP_006229893.1 | 9 | 1910 | |||
| XM_006229832.2 | XP_006229894.1 | 9 | 1898 | |||
| XM_006229834.2 | XP_006229896.1 | 8 | 1866 | |||
| XM_006229835.2 | XP_006229897.1 | 6 | 2249 | |||
| XM_006229837.2 | XP_006229899.1 | 8 | 1681 | |||
| XM_006229838.2 | XP_006229900.1 | 8 | 1669 | |||
| XM_008759662.1 | XP_008757884.1 | 6 | 1649 | |||
| XM_008759663.1 | XP_008757885.1 | 6 | 2190 | |||
| GenBank mRNA | AB236144.1 | - | 7 | 1156 | ||
| FQ230423.1 | - | 2 | 385 | |||
| U57715.1 | - | 8 | 1660 | |||

Pgap2
Genomic Map