siRNA Details
| Gene Symbol: Cgrrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053899.1 | NP_446351.1 | 2 | 274 | ||
| GenBank mRNA | BC078763.1 | - | 2 | 290 | 7107668 | |
| FQ218613.1 | - | 2 | 289 | |||
| U66471.1 | - | 2 | 274 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cgrrf1| Gene Symbol: Cgrrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053899.1 | NP_446351.1 | 2 | 274 | ||
| GenBank mRNA | BC078763.1 | - | 2 | 290 | 7107668 | |
| FQ218613.1 | - | 2 | 289 | |||
| U66471.1 | - | 2 | 274 | |||