siRNA Details
| Gene Symbol: Efna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053903.1 | NP_446355.1 | 3 | 505 | ||
| XM_006245593.2 | XP_006245655.1 | 3 | 698 | |||
| GenBank mRNA | U69279.1 | - | 3 | 505 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efna5| Gene Symbol: Efna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053903.1 | NP_446355.1 | 3 | 505 | ||
| XM_006245593.2 | XP_006245655.1 | 3 | 698 | |||
| GenBank mRNA | U69279.1 | - | 3 | 505 | ||