siRNA Details
| Gene Symbol: Inpp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053917.1 | NP_446369.1 | 25 | 4104 | ||
| XM_008772462.1 | XP_008770684.1 | 25 | 3948 | |||
| XM_008772465.1 | XP_008770687.1 | 24 | 3897 | |||
| GenBank mRNA | U96921.1 | - | 25 | 4104 | ||

Inpp4b
Genomic Map