siRNA Details
Gene Symbol: Sept5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053931.3 | NP_446383.3 | 2 | 360 | ||
GenBank mRNA | AB027143.1 | - | 2 | 360 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sept5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053931.3 | NP_446383.3 | 2 | 360 | ||
GenBank mRNA | AB027143.1 | - | 2 | 360 |