siRNA Details
| Gene Symbol: Eif2b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053950.1 | NP_446402.1 | 5, 6 | 501 | ||
| GenBank mRNA | Z48225.1 | - | 5, 6 | 501 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif2b4| Gene Symbol: Eif2b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053950.1 | NP_446402.1 | 5, 6 | 501 | ||
| GenBank mRNA | Z48225.1 | - | 5, 6 | 501 | ||