siRNA Details
| Gene Symbol: Mcf2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053951.2 | NP_446403.2 | 12 | 1475 | ||
| XM_006253416.2 | XP_006253478.1 | 12 | 4737 | |||
| XM_006253419.2 | XP_006253481.1 | 12 | 4737 | |||
| XM_006253421.2 | XP_006253483.1 | 12 | 4736 | |||
| XM_006253423.2 | XP_006253485.1 | 12 | 3363 | |||
| XM_006253424.2 | XP_006253486.1 | 12 | 3554 | |||
| XM_008771381.1 | XP_008769603.1 | 14 | 2705 | |||
| XM_008771382.1 | XP_008769604.1 | 12 | 2107 | |||
| XM_008771383.1 | XP_008769605.1 | 12 | 2390 | |||
| GenBank mRNA | Z35654.1 | - | 10 | 1568 | ||

Mcf2l
Genomic Map