siRNA Details
Gene Symbol: Ccr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053958.1 | NP_446410.1 | 3 | 951 | ||
XM_006244178.2 | XP_006244240.1 | 3 | 1729 | |||
GenBank mRNA | Y13400.1 | - | 3 | 951 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053958.1 | NP_446410.1 | 3 | 951 | ||
XM_006244178.2 | XP_006244240.1 | 3 | 1729 | |||
GenBank mRNA | Y13400.1 | - | 3 | 951 |