siRNA Details
| Gene Symbol: Nln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053970.2 | NP_446422.2 | 4 | 527 | ||
| XM_006231880.2 | XP_006231942.1 | 4 | 681 | |||
| XM_006231881.2 | XP_006231943.1 | 5 | 759 | |||
| GenBank mRNA | BC072687.1 | - | 4 | 527 | 7099208 | |
| X87157.1 | - | 4 | 635 | |||

Nln
Genomic Map