siRNA Details
| Gene Symbol: Nln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053970.2 | NP_446422.2 | 10 | 1684 | ||
| XM_006231880.2 | XP_006231942.1 | 10 | 1838 | |||
| XM_006231881.2 | XP_006231943.1 | 11 | 1916 | |||
| GenBank mRNA | BC072687.1 | - | 10 | 1684 | 7099208 | |
| FQ228111.1 | - | 3 | 391 | |||
| FQ230641.1 | - | 3 | 391 | |||
| X87157.1 | - | 10 | 1792 | |||

Nln
Genomic Map