siRNA Details
Gene Symbol: Pdha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053994.2 | NP_446446.1 | 1 | 366 | ||
GenBank mRNA | BC078757.1 | - | 1 | 366 | 7113421 | |
Z18878.1 | - | 1 | 419 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pdha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053994.2 | NP_446446.1 | 1 | 366 | ||
GenBank mRNA | BC078757.1 | - | 1 | 366 | 7113421 | |
Z18878.1 | - | 1 | 419 |