siRNA Details
| Gene Symbol: Pdha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053994.2 | NP_446446.1 | 1 | 370 | ||
| GenBank mRNA | BC078757.1 | - | 1 | 370 | 7113421 | |
| Z18878.1 | - | 1 | 423 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdha2| Gene Symbol: Pdha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053994.2 | NP_446446.1 | 1 | 370 | ||
| GenBank mRNA | BC078757.1 | - | 1 | 370 | 7113421 | |
| Z18878.1 | - | 1 | 423 | |||