siRNA Details
| Gene Symbol: Csde1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054006.1 | NP_446458.1 | 2 | 223 | ||
| GenBank mRNA | FQ223276.1 | - | 2 | 221 | ||
| X52311.1 | - | 2 | 223 | |||
| Gene Symbol: LOC100364335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751853.3 | XP_003751901.1 | 1 | 224 | ||

Csde1
Genomic Map