siRNA Details
| Gene Symbol: Lrrc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057142.1 | NP_476483.1 | 3 | 358 | ||
| GenBank mRNA | EU348640.1 | - | 2 | 281 | ||
| EU348641.1 | - | 2 | 282 | |||
| EU348642.1 | - | 3 | 358 | |||
| EU348643.1 | - | 3 | 358 | |||
| EU348644.1 | - | 3 | 358 | |||
| U66707.1 | - | 3 | 358 | |||

Lrrc7
Genomic Map