siRNA Details
| Gene Symbol: RNF24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134337.2 | NP_001127809.1 | 6 | 500 | ||
| NM_001134338.2 | NP_001127810.1 | 7 | 569 | |||
| NM_001321749.1 | NP_001308678.1 | 6 | 736 | |||
| NM_007219.4 | NP_009150.1 | 6 | 792 | |||
| XM_011529145.2 | XP_011527447.1 | 8 | 815 | |||
| XM_011529146.2 | XP_011527448.1 | 7 | 745 | |||
| XM_011529147.2 | XP_011527449.1 | 9 | 892 | |||
| XM_017027620.1 | XP_016883109.1 | 7 | 837 | |||
| XM_017027621.1 | XP_016883110.1 | 7 | 606 | |||
| XM_017027622.1 | XP_016883111.1 | 8 | 899 | |||
| GenBank mRNA | AL079313.1 | - | 4 | 208 | ||
| AL096778.1 | - | 6 | 590 | |||
| BC000213.2 | - | 7 | 523 | 2958640 | ||
| BC039584.1 | - | 6 | 419 | 5214313 | ||
| BT007406.1 | - | 5 | 338 | |||
| CV811405.1 | - | 6 | 478 | |||

RNF24
Genomic Map