siRNA Details
| Gene Symbol: Ncr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057199.1 | NP_476547.1 | 4 | 513 | ||
| GenBank mRNA | AF082533.1 | - | 4 | 513 | ||
| AJ012741.1 | - | 4 | 503 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncr1| Gene Symbol: Ncr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057199.1 | NP_476547.1 | 4 | 513 | ||
| GenBank mRNA | AF082533.1 | - | 4 | 513 | ||
| AJ012741.1 | - | 4 | 503 | |||