siRNA Details
| Gene Symbol: Kif1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057200.2 | NP_476548.1 | 46 | 5406 | ||
| XM_008764204.1 | XP_008762426.1 | 48 | 5478 | |||
| XM_008764205.1 | XP_008762427.1 | 46 | 5182 | |||
| GenBank mRNA | AB070355.1 | - | 46 | 5406 | ||
| AB120429.1 | - | 42 | 4665 | |||

Kif1b
Genomic Map