siRNA Details
| Gene Symbol: Ddit4l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080399.1 | NP_536324.1 | 773 | |||
| GenBank mRNA | AF327511.1 | - | 773 | |||
| Gene Symbol: Ddit4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729121.4 | XP_002729167.1 | 3 | 620 | ||

Ddit4l2
Genomic Map