siRNA Details
Gene Symbol: Nploc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080577.2 | NP_542144.1 | 8 | 899 | ||
GenBank mRNA | AF234600.1 | - | 8 | 827 | ||
BC101887.1 | - | 8 | 899 | 7930738 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nploc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080577.2 | NP_542144.1 | 8 | 899 | ||
GenBank mRNA | AF234600.1 | - | 8 | 827 | ||
BC101887.1 | - | 8 | 899 | 7930738 |