siRNA Details
| Gene Symbol: Manea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237960.1 | XP_006238022.1 | 5 | 2028 | ||
| XM_006237961.2 | XP_006238023.1 | 5 | 1947 | |||
| GenBank mRNA | FQ223109.1 | - | 5 | 1953 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Manea| Gene Symbol: Manea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237960.1 | XP_006238022.1 | 5 | 2028 | ||
| XM_006237961.2 | XP_006238023.1 | 5 | 1947 | |||
| GenBank mRNA | FQ223109.1 | - | 5 | 1953 | ||