siRNA Details
| Gene Symbol: Pde11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127480.1 | NP_001120952.1 | 22 | 3128 | ||
| NM_001127481.2 | NP_001120953.1 | 20 | 2732 | |||
| NM_080893.1 | NP_543169.1 | 20 | 3806 | |||
| GenBank mRNA | AB059360.1 | - | 20 | 2734 | ||
| AB059361.1 | - | 22 | 3128 | |||
| AB059362.1 | - | 20 | 3806 | |||

Pde11a
Genomic Map