siRNA Details
| Gene Symbol: Hnrnph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080896.1 | NP_543172.1 | 4 | 524 | ||
| GenBank mRNA | AF367468.1 | - | 4 | 524 | ||
| BC099792.1 | - | 603 | ||||
| BC129087.1 | - | 742 | 8363754 | |||
| FQ212957.1 | - | 1 | 103 | |||

Hnrnph1
Genomic Map