siRNA Details
| Gene Symbol: Bnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080897.1 | NP_543173.1 | 3 | 229 | ||
| GenBank mRNA | AF454391.1 | - | 3 | 229 | ||
| FQ218385.1 | - | 3 | 233 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bnip1| Gene Symbol: Bnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080897.1 | NP_543173.1 | 3 | 229 | ||
| GenBank mRNA | AF454391.1 | - | 3 | 229 | ||
| FQ218385.1 | - | 3 | 233 | |||