siRNA Details
| Gene Symbol: Higd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080902.4 | NP_543178.2 | 3 | 247 | ||
| GenBank mRNA | AY062253.3 | - | 3 | 306 | ||
| BC059118.1 | - | 3 | 251 | 6890532 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Higd1a| Gene Symbol: Higd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080902.4 | NP_543178.2 | 3 | 247 | ||
| GenBank mRNA | AY062253.3 | - | 3 | 306 | ||
| BC059118.1 | - | 3 | 251 | 6890532 | ||