siRNA Details
| Gene Symbol: Ddit4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080906.2 | NP_543182.1 | 3 | 1409 | ||
| GenBank mRNA | AF334162.1 | - | 3 | 1414 | ||
| BC062021.1 | - | 3 | 1342 | 5623446 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddit4| Gene Symbol: Ddit4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080906.2 | NP_543182.1 | 3 | 1409 | ||
| GenBank mRNA | AF334162.1 | - | 3 | 1414 | ||
| BC062021.1 | - | 3 | 1342 | 5623446 | ||