siRNA Details
| Gene Symbol: Slc36a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130415.1 | NP_569099.1 | 3 | 311 | ||
| XM_008767657.1 | XP_008765879.1 | 2 | 2410 | |||
| XM_008767659.1 | XP_008765881.1 | 4 | 993 | |||
| GenBank mRNA | AF361239.1 | - | 3 | 311 | ||

Slc36a1
Genomic Map