siRNA Details
Gene Symbol: Dcbld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130419.1 | NP_569103.1 | 4, 5 | 623 | ||
GenBank mRNA | AF387549.1 | - | 4, 5 | 623 |

More Information
Other Species Targeted

Gene Ontology Categories:
