siRNA Details
| Gene Symbol: Acot12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130747.1 | NP_570103.1 | 15 | 1640 | ||
| XM_008760628.1 | XP_008758850.1 | 16 | 2135 | |||
| GenBank mRNA | AB040609.1 | - | 15 | 1640 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acot12| Gene Symbol: Acot12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130747.1 | NP_570103.1 | 15 | 1640 | ||
| XM_008760628.1 | XP_008758850.1 | 16 | 2135 | |||
| GenBank mRNA | AB040609.1 | - | 15 | 1640 | ||