siRNA Details
| Gene Symbol: Hadha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130826.2 | NP_570839.2 | 5 | 370 | ||
| GenBank mRNA | BC091697.1 | - | 5 | 370 | 7301707 | |
| D16478.1 | - | 5 | 368 | |||
| X98225.1 | - | 3 | 256 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hadha| Gene Symbol: Hadha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130826.2 | NP_570839.2 | 5 | 370 | ||
| GenBank mRNA | BC091697.1 | - | 5 | 370 | 7301707 | |
| D16478.1 | - | 5 | 368 | |||
| X98225.1 | - | 3 | 256 | |||