siRNA Details
| Gene Symbol: Fgf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131908.1 | NP_571983.1 | 3 | 473 | ||
| GenBank mRNA | AB079674.1 | - | 3 | 473 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf6| Gene Symbol: Fgf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131908.1 | NP_571983.1 | 3 | 473 | ||
| GenBank mRNA | AB079674.1 | - | 3 | 473 | ||