siRNA Details
| Gene Symbol: Dynlrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131910.3 | NP_571985.1 | 3 | 321 | ||
| GenBank mRNA | AF073839.2 | - | 3 | 244 | ||
| AY026512.1 | - | 3 | 321 | |||
| BC058437.1 | - | 3 | 262 | 6921076 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dynlrb1| Gene Symbol: Dynlrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131910.3 | NP_571985.1 | 3 | 321 | ||
| GenBank mRNA | AF073839.2 | - | 3 | 244 | ||
| AY026512.1 | - | 3 | 321 | |||
| BC058437.1 | - | 3 | 262 | 6921076 | ||