siRNA Details
| Gene Symbol: Hemgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133294.1 | NP_579828.1 | 3 | 381 | ||
| XM_008763693.1 | XP_008761915.1 | 3 | 457 | |||
| XM_008763694.1 | XP_008761916.1 | 4 | 525 | |||
| GenBank mRNA | AJ302650.2 | - | 3 | 381 | ||
| BC078739.1 | - | 4 | 481 | 7111835 | ||

Hemgn
Genomic Map