siRNA Details
| Gene Symbol: Hemgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133294.1 | NP_579828.1 | 4 | 1921 | ||
| XM_008763693.1 | XP_008761915.1 | 4 | 2005 | |||
| XM_008763694.1 | XP_008761916.1 | 5 | 2073 | |||
| GenBank mRNA | AJ302650.2 | - | 4 | 1921 | ||
| FQ231115.1 | - | 2 | 677 | |||
| FQ232586.1 | - | 2 | 706 | |||

Hemgn
Genomic Map